Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Thermometer Clamp
SDPSCI_ED

Catalog No. S15769 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Color:
Silver
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
S15769 Silver
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S15769 Supplier United Scientific™ Supplier No. THCL01
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sturdy Clamp that can be used with virtually all support stands and lattice setups.

  • Thermometer Clamp features an adjustable 360° rotating jaw for suspending thermometers.
  • The clamp attaches to rods up to 13 mm O.D.
  • Clamp can be locked at any angle.

Specifications

Quantity 1 Ea.
Type Thermometer Clamp
Material Aluminum
Color Silver
Jaw Type Spring Loaded
Diameter (Metric) Inside 13 mm

Warning: Cancer and reproductive harm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.