Learn More
MilliporeSigma™ Supelco™ Thermometer L 5in., temperature -10-110 C
Catalog No. 111010985 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Material:
Glass
Catalog No. Material
11-101-0985 Glass
1 options
Catalog No. 11-101-0985 Supplier MilliporeSigma™ Supelco™ Supplier No. 57332
Add to Cart
Add to Cart
For monitoring sample temperature when using the SPME sampling stand and a heat/stir plate.
TRUSTED_SUSTAINABILITY
Specifications
Material Glass