Learn More
AdipoGen Thermorubin
SDP
Supplier:  AdipoGen AGCN20339M001
Encompass_Preferred
Chemical. CAS 37577-75-6. Formula C32H24O12. MW 600.5. Isolated from Thermoactinomyces sp. Anthracene antibiotic. Antibacterial agent. Active against both Gram-positive and Gram-negative bacteria. Inhibits the initiation stage of bacterial protein synthesis by binding to the inter-subunit bridge B2a on the 70S ribosome. Does not inhibit DNA and RNA synthesis. Potent aldose reductase inhibitor.
Catalog No. 50-256-2565
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.