Learn More
Sonoco™ ThermoSafe Insulated EPS with Corrugate Containers and Coolers (Large)
Large selection of cost-effective, reusable EPS foam in corrugate stock containers available in small, medium, and large sizes. EPS foam with corrugate shippers for the safe storage and delivery of most refrigerated or frozen products.
Supplier: Sonoco™ 638
Description
- Sizes that range 19 inches- 24 inches in length
- Also available in ThermoSafe Insulated EPS with Corrguate Containers and Coolers- Small (6 inches-10.5 inches in length)
- Also available in ThermoSafe Insulated EPS with Corrguate Containers and Coolers- Medium (11 inches-17 inches in length)
- 200 lb. test corrugated
- Excellent dry ice sublimation… average dry ice usage in 1-1/2 inch thick wall units is 5 pounds over 24 hours and 3-4 pounds thereafter.
- Standard 1-1/2 inch wall shippers are ideal for refrigerated shipments for 24-48 hours.
Specifications
2 per carton | |
1.125 in. | |
Expanded Polystyrene Insulated |
28.25 X 17.25 X 15.63 in. | |
24 X 13.88 X 12.5 in. |