Learn More
Sonoco™ ThermoSafe LabMailer Specimen Tube Mailers
Shock-absorbing polystyrene foam mailers accept sample tubes with rubber or screw caps
$175.90 - $471.50
Specifications
Applications | Ideal for mailing or transporting nonhazardous blood, urine, or other lab samples |
---|---|
Material | Polystyrene Foam |
Catalog Number | Mfr. No. | Capacity | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity | Quantity | Price | Quantity & Availability | ||||
03-525-22A
|
Sonoco™
362 |
5, 7, or 10 mL | 2 Tubes |
Pack of 150 for $307.00
|
|
||||
03-527
![]() |
Sonoco™
356 |
3, 5, 7 or 10 mL | 1 Tube |
Pack of 300 for $471.50
|
|
||||
03-528
![]() |
Sonoco™
364 |
3, 5, 7 or 10 mL | 3 Tubes |
Pack of 150 for $277.50
|
|
||||
03-530-35
![]() |
Sonoco™
409 |
7, 10, 15 or 25 mL | 5 Tubes |
Pack of 100 for $356.00
|
|
||||
03-530-5
|
Sonoco™
341 |
5, 7 or 10 mL | 8 Tubes |
Pack of 50 for $175.90
|
|
||||
03-531-20
|
Sonoco™
339 |
3 or 5 mL | 5 Tubes |
Pack of 150 for $281.00
|
|
||||
03-600-165
![]() |
Sonoco™
407 |
7 to 10 mL | 4 Tubes |
Pack of 100 for $262.50
|
|
||||
Description
- Two-piece assembly with snap-lock fasteners holds contents in place
- Ideal for mailing or transporting nonhazardous blood, urine, or other lab samples.
†Includes slotted area for up to four slides.
Specifications
Ideal for mailing or transporting nonhazardous blood, urine, or other lab samples | |
Polystyrene Foam |