Description
Includes
24 oz. Polar pack tough pack (8), 20 oz. Polar pack standard gel pack (3), corrugated outer corepack (1 or 2), double-wall laminated pads (1), corrugated inner corepack (1)
- Meet toughest ambient profiles of any premium shipper in the industry
- Multicomponent packing system to ensure exceptional temperature protection and durability
- Model E48UPS:
- Complies with international clean air regulations, CFC- and HCFC-free
- Qualified for maintaining payloads of a single 1mL vial to 3L of liquid
- Reusable
- Replaceable components available separately
- With two double-wall laminated pads
- Model E24AUPS:
- Specifically engineered for quick trips of 24 hours or less
- Lightweight and cost-effective EPS inner shell reduces overnight shipping costs
- Superior insulation power of 2 in. thick walls
- With one double-wall laminated pad
Order Info
Container, corepacks and gels shipped in a single overpack.
Specifications
Specifications
| Applications | For refrigerated, frozen pharmaceutical and biological specimens |
| Type | Outer Corepack |