Learn More
New England Biolabs, Inc. Thermostable Inorganic Pyrophosphatase – 1250 units
Supplier: New England Biolabs, Inc. M0296L
Inorganic pyrophosphatase (PPase) catalyzes the hydrolysis of inorganic pyrophosphate to form orthophosphate: P2O7-4 + H2O -> 2HPO4-2.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.