Learn More
Medchemexpress LLC Thiabendazole-d4 | 1190007-20-5 | 98.8% | 205.27 g/mol | C10H3D4N3S | 10 MG
Supplier: Medchemexpress LLC HYB0263S10MG
Thiabendazole-d4 is the deuterated isotopologue of thiabendazole, supplied as a solid stable isotope-labeled reference material for analytical and research use. It is formulated for use as an internal standard and reference in mass spectrometry and quantitative assays.
- Deuterated internal standard for mass spectrometry and quantitative assays.
- High chemical purity (98.84%) for reliable analytical performance.
- White to off-white solid, suitable for handling and weighing.
- Available in small, accurately weighed sizes for reference and calibration.
- Powder stable at -20°C for long-term storage; solvent stability is limited and should follow recommended conditions.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.