Learn More
Thermo Scientific Chemicals Thiacloprid
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 133.0°C to 136.0°C |
| Quantity | 1 g |
| Solubility Information | Solubility aq. soln.: .. Other solubilities: insoluble in chloroform & methanol |
| Formula Weight | 252.72 |
| Chemical Name or Material | Thiacloprid |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Toxic if swallowed. Harmful if inhaled. May cause drowsiness or dizziness. Suspected of causing cancer. May damage fertility. May damage the unborn child. Very toxic to aquatic life with long last |
| Precautionary Statement | GHS P Statement Wash face, hands and any exposed skin thoroughly after handling. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Wear protective gloves/protective clothing/eye protection/face protection. |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.