Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thiamethoxam, Thermo Scientific™

Catalog No. AC470050010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC470050010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AC470050010 Supplier Thermo Scientific Chemicals Supplier No. 470050010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
This item is not returnable. View return policy

Specifications

Chemical Name or Material Thiamethoxam
CAS 153719-23-4
Synonym 3-(2-Chloro-5-thiazolylmethyl)tetrahydro-5-methyl-N-nitro-4 H-1,3,5-oxadiazin-4-imine
Quantity 1 g
Melting Point 137°C to 143°C
Shipping Condition No special requirements
Product Line Life science
Molecular Formula C8 H10 Cl N5 O3 S
Physical Form Powder or Crystals
Color White to Light-yellow to Light-beige
Purity ≥97.5%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.