Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thiamine Mononitrate, FCC, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18607365 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
25 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-365 25 kg Amber Glass Bottle
18-607-361 25 g Amber Glass Bottle
18-607-362 100 g Amber Glass Bottle
18-607-363 500 g Amber Glass Bottle
18-607-364 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-607-365 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. T105425KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

T1054, 532-43-4, C12H17N5O4S

Spectrum™ Chemical Thiamine Mononitrate, FCC is in the B vitamin group and is water soluble. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 532-43-4
Molecular Formula C12H17N5O4S
Molecular Weight (g/mol) 327.36
MDL Number MFCD00036330
InChI Key UIERGBJEBXXIGO-UHFFFAOYSA-N
IUPAC Name 3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-5-(2-hydroxyethyl)-4-methyl-3H-1λ4,3-thiazol-1-ylium nitrate
SMILES [O-][N+]([O-])=O.CC1=C(CCO)[S+]=CN1CC1=CN=C(C)N=C1N

Specifications

CAS Max % 1
pH 6.0 to 7.5
Ignition Residue 0.002
Loss on Drying 0.01
Packaging Amber Glass Bottle
Quantity 25 kg
Percent Purity 98 to 102%
Grade FCC
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Thiamine Mononitrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.