Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Thiazolyl Blue Tetrazolium Bromide, Membrane-Permeable, >98% Purity, 500 mg

Catalog No. 502583230
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-3230 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-3230 Supplier Abcam Supplier No. AB146345500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

High-purity, membrane-permeable dye for biochemical analysis.

  • Molecular Weight: 414.3 Da
  • Purity: >98%
  • Form: Solid
  • CAS Number: 298-93-1
  • Storage Temperature: Store at +4°C
  • Shipping Temperature: Blue Ice
  • Citations: 20

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Substrate Properties Chemical Substrate
Substrate MTT
Shipping Condition Blue Ice
Buffer 100% thiazolyl blue tetrazolium bromide
Content And Storage Store at 4°C.
Form Solid
Detection Method Colorimetric
Formulation 100% Thiazolyl blue tetrozolium bromide
Quantity 500 mg
For Use With (Application) Sample Prep and Detection
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.