Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Thiazolyl Blue Tetrazolium Bromide, Solid, 414.3 Da, 5 g

Catalog No. 502583229
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-3229 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-3229 Supplier Abcam Supplier No. AB1463455G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity, membrane-permeable dye for biochemical analysis.

  • Molecular weight: 414.3 Da
  • Purity: >98%
  • Form: Solid
  • Storage: Store at +4°C
  • Shipping: Blue ice
  • Composition: 100% Thiazolyl blue tetrozolium bromide
  • CAS: 298-93-1
  • Citations: 20

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Thiazolyl Blue Tetrazolium Bromide
Substrate MTT
Shipping Condition Blue Ice
Substrate Properties Chemical Substrate
Buffer 100% thiazolyl blue tetrazolium bromide
Content And Storage Store at 4°C.
Alternate Part Number 298-93-1
Form Solid
Detection Method Colorimetric
Formulation 100% Thiazolyl blue tetrazolium bromide
Quantity 5 g
For Use With (Application) Sample Prep and Detection
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.