Learn More
Abcam Thiazolyl Blue Tetrazolium Bromide, Solid, 98% Purity, 1 g
Description
- Molecular weight: 414.3 Da
- Purity: >98%
- Form: Solid
- Composition: 100% Thiazolyl blue tetrozolium bromide
- Storage temperature: Store at +4°C
- Shipping temperature: Blue Ice
- CAS Number: 298-93-1
- Citations: 20
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Substrate Properties | Chemical Substrate |
| Substrate | MTT |
| Shipping Condition | Blue Ice |
| Content And Storage | Store at 4°C. |
| Detection Method | Colorimetric |
| Buffer | 100% Thiazolyl blue tetrazolium bromide |
| Form | Solid |
| Formulation | 100% Thiazolyl blue tetrozolium bromide |
| Quantity | 1 g |
| For Use With (Application) | Functional studies |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.