Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eisco™ Thick Walled Mortar and Pestle Set

Eisco™ Thick Walled Mortar and Pestle Set is made from lab quality porcelain and feature a thick wall design, flat bottom, unglazed grinding surfaces for efficient grinding, and spout for easy pouring.
Supplier: Eisco™ CH0541B
Description
- Made from lab quality porcelain
- Thick wall design
- Outside of mortar and handle of pestle is glazed
- Inner bowl and pestle end are not glazed
- Spout for easing pouring
- Flat bottom for stability
Specifications
70 mL | |
Autoclavable | |
Mortar and Pestle | |
Grinding | |
Thick Walled |
Glazed Outer | |
8.2 cm | |
Porcelain | |
Mortar, Pestle |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction