Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thieno[3,2-b]thiophene-2-carbonitrile 98.0+%, TCI America™
SDP

Catalog No. T32211G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T32211G 1 g
T3221200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T32211G Supplier TCI America Supplier No. T32211G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 40985-58-8
Molecular Formula C7H3NS2
Molecular Weight (g/mol) 165.228
MDL Number MFCD08059461
InChI Key UPRBPGCXCNBTQH-UHFFFAOYSA-N
Synonym 2-Cyanothieno[3,2-b]thiophene
PubChem CID 55252708
IUPAC Name thieno[3,2-b]thiophene-5-carbonitrile
SMILES C1=CSC2=C1SC(=C2)C#N

Specifications

Melting Point 50°C
Color White-Yellow
Quantity 1 g
Formula Weight 165.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Thieno[3,2-b]thiophene-2-carbonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.