Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thifluzamide 98.0+%, TCI America™
SDP

Catalog No. T36481G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T36481G 1 g
T3648250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T36481G Supplier TCI America Supplier No. T36481G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 130000-40-7
Molecular Formula C13H6Br2F6N2O2S
Molecular Weight (g/mol) 528.061
MDL Number MFCD03428071
InChI Key WOSNCVAPUOFXEH-UHFFFAOYSA-N
Synonym N-[2,6-Dibromo-4-(trifluoromethoxy)phenyl]-2-methyl-4-(trifluoromethyl)thiazole-5-carboxamide
PubChem CID 86389
ChEBI CHEBI:81794
IUPAC Name N-[2,6-dibromo-4-(trifluoromethoxy)phenyl]-2-methyl-4-(trifluoromethyl)-1,3-thiazole-5-carboxamide
SMILES CC1=NC(=C(S1)C(=O)NC2=C(C=C(C=C2Br)OC(F)(F)F)Br)C(F)(F)F

Specifications

Melting Point 177°C
Quantity 1 g
UN Number 3077
Formula Weight 528.06
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Thifluzamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.