Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thimerosal, Fisher BioReagents

Catalog No. BP254225 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Amber Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP254225 25 g Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Catalog No. BP254225

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
  • Thimerosal is a mercury-containing compound commonly used as an antibacterial and antifungal agent
  • Also used as a bactericide in pharmaceuticals and research
  • Intended for research use only
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54-64-8
Molecular Formula C9H9HgNaO2S
Molecular Weight (g/mol) 404.81
MDL Number MFCD00013062
InChI Key RTKIYNMVFMVABJ-UHFFFAOYSA-L
Synonym Sodium ethylmercurithiosalicylate
PubChem CID 16684434
ChEBI CHEBI:9546
IUPAC Name sodium;(2-carboxylatophenyl)sulfanyl-ethylmercury
SMILES [Na+].CC[Hg]SC1=CC=CC=C1C([O-])=O

Specifications

Type Thimerosal
Physical Form Solid
pH 6.7 to 8.2
Melting Point 232°C
Loss on Drying 0.5% max.
Packaging Amber Glass
Color Cream
Quantity 25 g
Assay Percent Range ≥97 %
Merck Index 15, 9470
Formula Weight 404.8
Percent Purity 97% min.
Chemical Name or Material Thimerosal
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
May be fatal if inhaled, absorbed through skin, or swallowed. Danger of cumulative effects. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use only under a chemical fume hood. Use personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required.

NFPA

Health:4
Flammability:1
Instability:0
Recommended Storage RT, protect from light, desiccate.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.