Description
- Clear polystyrene housing with sealed PET capillary pore membrane
- Hanging geometry provides protection to lower compartment cell layer
- Simplified pipetting due to self-lift geometry
- Eccentric in-well positioning allows unequalled access to lower compartment
- Available in 6, 12 and 24 well format
Specifications
Specifications
| Material | Polystyrene housing with sealed PET capillary pore membrane |
| Pore Density | 2 x 106 pores/cm2 |
| Pore Size | 3 μm |
| No. of Wells | 24 |