Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thioacetamide, CP, Spectrum™ Chemical
SDP

Catalog No. 18614382 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
500 g
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-614-382 25 kg Fiber Drum
18-614-381 25 g Amber Glass Bottle
18-614-383 500 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-614-382 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. T104225KGBL
Add to Cart
Add to Cart
Request a Quote

Chemical Identifiers

CAS 62-55-5

Specifications

CAS Max % 1
Packaging Fiber Drum
Quantity 25 kg
Grade Chemically Pure
WARNING: Cancer - www.P65Warnings.ca.gov

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.