Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thioacetamide TS, 4% w/v, Reagents

Catalog No. 01333458
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-333-458 500 mL
01-333-459 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-333-458 Supplier Reagents Holdings Llc Supplier No. CT010400500C
Add to Cart
Add to Cart

Thioacetamide TS, 4% w/v, Meets USP specifications, for general laboratory use. Recommended Storage: 2°C to 8°C (36°F to 46°F) Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 62-55-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Thiocarbamoyl Hydrazide, Aminothiourea, Hydrazinecarbothioamide
IUPAC Name water
SMILES O

Specifications

Color Colorless to Slight Yellow
Boiling Point 100°C
Packaging Glass Bottle
Quantity 500 mL
Specific Gravity 1.02
Percent Purity 4%
Grade USP
Physical Form Liquid
Chemical Name or Material Thioacetamide TS, 4% w/v
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.