Learn More
Sigma Organic Chemistry Thioglycolic acid | 500ML | 68-11-1 | MFCD00004876
Supplier: Sigma Organic Chemistry T3758500ML

Thioglycolic acid, 500ML
About This Item:
Synonym(s): Mercaptoacetic acid
Linear Formula: HSCH2COOH
Molecular Weight: 92.12
CAS Number: 68-11-1
MDL Number: MFCD00004876
UNSPSC Code: 12352100
Purity: 0.98
Boiling Point: 96 C/5 mmHg (lit.)
Melting Point: -16 C (lit.)
Density: 1.326 g/mL at 20 C (lit.)
Refractive Index: n20/D 1.505 (lit.)
Storage: 2-8C
EINECS Number: 200-677-4
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.