Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Thiol Green Indicator, Solid, 5 mg

Catalog No. 502704476
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-4476 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-4476 Supplier Abcam Supplier No. AB1560495MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Fluorescent indicator designed for the detection of thiols in biological samples.

  • Form: Solid
  • Constituents: 100% Green Indicator

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Kit Contents 5 mg Green Indicator
Substrate Properties Chemical Substrate
Target Molecule Thiols
Label or Dye Thiol Green
Buffer 100% Green Indicator
Form Solid
Detection Method Fluorescence
Formulation 100% Green Indicator
Quantity 5 mg
For Use With (Application) Functional studies
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.