Learn More
Invitrogen™ ThiolTracker™ Violet (glutathione detection reagent)
Description
ThiolTracker™ Violet offers the following;
Subcellular detection and localization of GSH is important in understanding the modulation of redox status, the effect of drugs, and the mechanisms of detoxification. Furthermore, differences in GSH levels in response to oxidative stress in subpopulations of cells have been reported, underscoring the importance of detection methods amenable to flow cytometry and automated fluorescence detection. ThiolTracker™ Violet can be used for Flow Cytometry, HCS imaging, and epifluorescent microscopy.
Specifications
Specifications
| Color | Violet |
| Content And Storage | Store at ≤–20°C, L protect material from long-term exposure to light; may be exposed to light for short periods of time, requires DMSO |
| Excitation Wavelength Range | 405⁄526 |
| Dye Type | ThiolTracker™ Violet |
| Format | Tube(s), Slide(s) |
| For Use With (Equipment) | Fluorescence Microscope, Flow Cytometer |
| Quantity | 180 reactions |
| Detection Method | Fluorescence |
| Emission | Visible |
| Form | Lyophilized |
| Show More |
Frequently Asked Questions (FAQs)
No. ThiolTracker Violet is fluorescent whether bound or unbound to a sulfhydryl group.
Yes, ThiolTracker Violet will bind to any accessible sulfhydryl group (-SH; cysteine) whether it is on reduced glutathione or on the surface of a protein. It will not bind to sulfur bound via a disulfide bond (cystine) or ‘buried’ cysteines.
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.