Learn More
Lkt Laboratories CAS 87-11-6, ≥98%, Bacterial and fungal RNA polymerase inhibitor. It prevents synthesis of RNA and degradation of mRNA.
SDP
Supplier:  Lkt Laboratories T2834
Encompass_Preferred
Thiolutin is a dithiolopyrrolone antibiotic synthesized by Streptomyces. Thiolutin displays broad spectrum antibacterial activity as well as some antifungal and anti-angiogenic activities. Thiolutin inhibits bacterial and yeast RNA polymerase, preventing RNA synthesis or transcription. Thiolutin also inhibits mRNA degradation. In vitro, this compound modulates HSP27 interactions to decrease actin stress fiber and F-actin levels and to increase cortical actin levels, resulting in inhibition of endothelial cell adhesion. Additionally, thiolutin decreases levels of paxillin and prevents endothelial cell adhesion in other in vitro models, inhibiting angiogenesis in S-180 tumor cells.
Catalog No. NC2539858
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.