Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thiomersal, BP, EP, 97-101%, Spectrum™ Chemical
SDP

Catalog No. 18607377 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-377 25 g Amber Glass Bottle
18-607-378 100 g Amber Glass Bottle
18-607-379 500 g Amber Glass Bottle
18-607-380 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-607-377 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. T171825GM
Add to Cart
Add to Cart

T1718, 54-64-8, C9H9HgNaO2S

Spectrum™ Chemical Thiomersal, BP, EP

Chemical Identifiers

CAS 54-64-8
Molecular Formula C9H9HgNaO2S
Molecular Weight (g/mol) 404.81
MDL Number MFCD00013062
InChI Key RTKIYNMVFMVABJ-UHFFFAOYSA-L
IUPAC Name sodium [(2-carboxylatophenyl)sulfanyl](ethyl)mercury
SMILES [Na+].CC[Hg]SC1=CC=CC=C1C([O-])=O

Specifications

Physical Form Crystalline Powder
pH 6.0 to 8.0
CAS Max % 1
Loss on Drying 0.005
Packaging Amber Glass Bottle
Sterility Non-sterile
Color White or White/Yellow
Quantity 25 g
Percent Purity 97 to 101%
Grade Multi-Compendial
Chemical Name or Material Thiomersal
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.