Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Thiophene-2-boronic acid, 98%
Click to view available options
Quantity:
1g
5g
25g
Description
Applications
Thiophene-2-boronic acid is a reagent used for Palladium-catalyzed Suzuki-Miyaura cross-couplings, Copper-catalyzed nitration reactions, Chain-growth catalyst transfer polycondensation of conjugated alternating copolymer, Ferric perchlorate-promoted reaction of fullerene to give fullerenyl boronic esters. It is also used in the preparation of Amino pyridine-based inhibitors of mitotic kinase Nek2 with potential antipoliferative effects in cancer tumors.
Solubility
Soluble in methanol.
Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
Thiophene-2-boronic acid is a reagent used for Palladium-catalyzed Suzuki-Miyaura cross-couplings, Copper-catalyzed nitration reactions, Chain-growth catalyst transfer polycondensation of conjugated alternating copolymer, Ferric perchlorate-promoted reaction of fullerene to give fullerenyl boronic esters. It is also used in the preparation of Amino pyridine-based inhibitors of mitotic kinase Nek2 with potential antipoliferative effects in cancer tumors.
Solubility
Soluble in methanol.
Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
Specifications
Quantity | 25g |
Melting Point | 128°C to 131°C |
Beilstein | 112375 |
Solubility Information | Soluble in methanol. |
Formula Weight | 127.96 |
Percent Purity | 98% |
Chemical Name or Material | Thiophene-2-boronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Keep cold