Learn More
Medchemexpress LLC Thiotaurine | 2937-54-4 | 99.3% | 141.21 g/mol | C2H7NO2S2 | 10 MG

Supplier: Medchemexpress LLC HYN1515610MG
Thiotaurine is an endogenous sulfur-containing metabolite of cystine provided as a high-purity research reagent. It acts as a sulfur donor, has been reported to modulate GAPDH activity, and may participate in neutrophil activation and leukocyte energy metabolism; it is described as a potential anti-inflammatory and immunomodulatory agent.
- High purity (99.27%).
- Molecular weight 141.21 g/mol.
- Chemical formula C2H7NO2S2.
- Supplied in milligram-scale quantities for laboratory research.
- Used for biochemical and cellular research applications.
- For research use only; not for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.