Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thiourea, Cytiva

Catalog No. 45001296 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-001-296 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-001-296 Supplier Cytiva Supplier No. RPN6301

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 45-001-296
Molecular Formula CH4N2S

Specifications

Color White
Quantity 100 g
Formula Weight 76.13
Physical Form Solid
Chemical Name or Material Thiourea
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.