Learn More
Medchemexpress LLC Thioxanthone | 492-22-8 | MFCD00005066 | >98.0% | 212.27 g/mol | C13H8OS | 100 G
SDP
Supplier:  Medchemexpress LLC HYW039724100G
Encompass_Preferred
Thioxanthone is a heterocyclic organic compound (IUPAC: thioxanthen-9-one; CAS 492-22-8) used as a photocatalyst and photoinitiator in organic synthesis and UV-curing applications. It is typically supplied as a yellow to light-yellow crystalline solid with high purity and participates in electron-transfer and energy-transfer processes, including interactions with metal complexes.
  • Heterocyclic photocatalyst suitable for photoredox reactions.
  • Effective photoinitiator for UV-curing coatings and inks.
  • Yellow to light-yellow crystalline solid for easy handling.
  • High purity (>98.0%) suitable for research applications.
  • Molecular weight 212.27 g/mol; chemical formula C13H8OS.
  • Compatible with metal-complex-mediated merger reactions.
Catalog No. 50-003-00394