Learn More
Medchemexpress LLC HY-10232 25mg Medchemexpress, THIP CAS:64603-91-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1023225MG
Encompass_Preferred
Medchemexpress, HY-10232 25mg THIP CAS:64603-91-4 THIP (Gaboxadol) is a selective δ-aminobutyric acid type A receptor (δ-GABAAR) agonist, functionally selective GABAAR ligand, exhibits agonism at α4β1δ, α4β3δ and weak antagonism at αβγ and α4β2δ GABAARs. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0652
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.