Learn More
Bachem Human Growth Hormone (1-43)
SDP
Supplier:  Bachem 4030633.1000
Encompass_Preferred
Human Growth Hormone (1-43), H-Phe-Pro-Thr-Ile-Pro-Leu-Ser-Arg-Leu-Phe-Asp-Asn-Ala-Met-Leu-Arg-Ala-His-Arg-Leu-His-Gln-Leu-Ala-Phe-Asp-Thr-Tyr-Gln-Glu-Phe-Glu-Glu-Ala-Tyr-Ile-Pro-Lys-Glu-Gln-Lys-Tyr-Ser-OH, 96827-07-5, C240H358N62O67S, Mr 5215.93, 1mg. Somatotropin (1-43) (human), hGH (1-43), Growth Hormone (1-43), human. This insulin-potentiating hGH-fragment has been detected in human serum. Treatment of obese yellow Avy/A mice with hGH (1-43) enhanced the in vitro sensitivity of their adipose tissue to insulin.
Catalog No. 50-194-7479
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.