Learn More
Bachem Galanin-Like Peptide (porcine) Trifluoroacetate
SDP
Supplier:  Bachem 4033157.0500
Encompass_Preferred
Galanin-Like Peptide (porcine) Trifluoroacetate, H-Ala-Pro-Val-His-Arg-Gly-Arg-Gly-Gly-Trp-Thr-Leu-Asn-Ser-Ala-Gly-Tyr-Leu-Leu-Gly-Pro-Val-Leu-His-Pro-Pro-Ser-Arg-Ala-Glu-Gly-Gly-Gly-Lys-Gly-Lys-Thr-Ala-Leu-Gly-Ile-Leu-Asp-Leu-Trp-Lys-Ala-Ile-Asp-Gly-Leu-Pro-Tyr-Pro-Gln-Ser-Gln-Leu-Ala-Ser-OH, 245114-96-9, C281H443N81O78, Mr 6204.11, 0.5mg. GALP (porcine). This member of the galanin family has been isolated from porcine hypothalamus. It is a 60 amino acid peptide that, unlike galanin, has a non-amidated C-terminus. GALP amino acid residues (9-21) are identical to the biologically active N-terminal (1-13) portion of galanin. Receptor binding studies revealed that GALP had a high affinity for the galanin receptor 2 (GALR2; IC50 = 0.24 nM) and a lower affinity for the GALR1 receptor (IC50 = 4.3 nM). GALP is therefore an endogenous ligand that preferentially binds the GALR2 receptor, whereas galanin is relatively non-selective.
Catalog No. 50-194-7475
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.