Learn More
Bachem Teplow's Amyloid Beta Protein (1-42) (scrambled II) Trifluoroacetate
SDP
Supplier:  Bachem 4104168.1000
Encompass_Preferred
Teplow's Amyloid Beta Protein (1-42) (scrambled II) Trifluoroacetate, H-Tyr-His-Ala-Gly-Val-Asp-Lys-Glu-Val-Val-Phe-Asp-Glu-Gly-Ala-Gly-Ala-Glu-His-Gly-Leu-Ala-Gln-Lys-Ile-Val-Arg-Gly-Phe-Gly-Val-Ser-Asp-Val-Ser-Met-Ile-His-Ile-Asn-Leu-Phe-OH, 1987844-92-7, C203H311N55O60S, Mr 4514.10, 1mg. b-Amyloid (1-42) (scrambled II), Ab42 scrambled II. This peptide is a specifically designed negative control in studies with Abeta42. It is "scrambled", which means it contains the same amino acids as Abeta42, but in different order. Referring to studies by Yamin and coworkers, Teplow's Amyloid b-Protein (1-42) does not show a number of phenomena regularly observed with Abeta42 (fibril formation, oligomerization, toxicity to neurons) and furthermore has a relatively flat hydropathy profile, which can be an advantage in several studies, for example in order to avoid unspecific interaction with the cell membrane.
Catalog No. 50-194-6593
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.