Learn More
ATCC THLE-3; Liver; Human (Homo sapiens)
Supplier:  ATCC CRL3583
Encompass
THLE-3 is an epithelial cell isolated from the left lobe of a donor. This cell line was deposited by National Cancer Institute and be used in toxicology research. This product is an ATCC manufactured and accessioned progeny of ATCC CRL-11233 cited in US Pat. No. 5,506,131.
Catalog No. NC2400619