Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ THOC7 Recombinant Protein Antigen
SDP

Catalog No. NBP182076PE Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP182076PE 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP182076PE Supplier Novus Biologicals™ Supplier No. NBP182076PEP
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human THOC7. The THOC7 Recombinant Protein Antigen is derived from E. coli. The THOC7 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.

This is a blocking peptide for NBP1-82076. This is a human recombinant protein expressed in E. coli with a N-terminal His-ABP tag and purified with IMAC chromatography.

Specifications

Gene ID (Entrez) 80145
Species Human
Purification Method Chromatography
Purity >80%
Concentration 0.5mg/mL
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Formulation PBS and 1M Urea, pH 7.4.
For Use With (Application) Blocking/Neutralizing, Control
Gene Symbol THOC7
Label Type Unlabeled
Molecular Weight (g/mol) 29kDa
Product Type THOC7
Quantity 0.1 mL
Regulatory Status RUO
Research Category Epigenetics
Source E.Coli
Specific Reactivity This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP1-82076. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml
Immunogen MGAVTDDEVIRKRLLIDGDGAGDDRRINLLVKSFIKWCNSGSQEEGYSQYQRMLSTLSQCEFSMGKTLLVYDMNLREMENYEKIYKEIECSIAGAHE
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.