Learn More
Thomson Instrument Company Thomson Filter Plate, 96 well, 2mL | 0.2um PVDF | CS20
SDP
Supplier:  Thomson Instrument Company 921731
Encompass_Preferred
Thomson Instrument Company filter plates are offered in both 24 and 96-well configurations, and designed for analytical and biological sample preparation. Depending on your application we may recommend using a positive pressure manifold, centrifugation or a high efficiency vacuum manifold that connects directly to your house vacuum.  The plates are designed to fit SLAS specifications and work well with automated liquid handling systems. PVDF, PTFE and PP filter plates are compatible with a wide variety of solvents.  Their long funnel design eliminates cross-contamination between sample collection wells by inserting below the top of the collection plate. Thomson Instrument Company manufactures its products in the USA.
Catalog No. 50-222-1350
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.