Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ThPOK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552351
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552351 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552351 Supplier Thermo Scientific Supplier No. PA552351
Only null left
Add to Cart
Add to Cart

ThPOK Polyclonal Antibody

ThPOK Polyclonal Antibody for ICC/IF, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen ThPOK
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Zbtb7b
Gene Accession No. O15156
Gene Alias CKROX; c-KROX; DKFZp686G01254; hcKROX; Krueppel-related zinc finger protein cKrox; RP11-307C12.9; T helper-inducing POZ/Krueppel factor; T-helper-inducing POZ/Krueppel-like factor; Thpok; Vzinc finger and BTB domain containing 7B; ZBTB15; ZBTB7B; ZFP67; Zfp-67; zinc finger and BTB domain containing 15; zinc finger and BTB domain containing 7B; Zinc finger and BTB domain-containing protein 15; Zinc finger and BTB domain-containing protein 7B; zinc finger protein 67; Zinc finger protein 67 homolog; zinc finger protein 857B; Zinc finger protein Th-POK; ZNF857B
Gene Symbols Zbtb7b
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZBTB7B. Recombinant protein control fragment (Product #RP-89376).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51043
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.