Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

THRAP3/TRAP150 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501558878
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-155-8878 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-155-8878 Supplier Bethyl Laboratories, Inc Supplier No. A300955A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse and X. laevis

THRAP3 is a 955 amino acid novel nuclear receptor and a member of the Thyroid hormone receptor associated proteins with a LISH domain and no obvious known sequence motifs. THRAP3 is a 150 kDa subunit of the large multiprotein complex TRAP and is important in the maintece of the fundamental structure of the fibrillar center and dense fibrillar component in the nucleolus. It is also known to play an important role in transcriptional coactivation. It is known to interact with DKC1, nop5/nap65 and fibrillarin. From the Northern blot analysis of several human tissues it was found that TRAP150 is ubiquitously expressed.

Specifications

Antigen THRAP3/TRAP150
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene THRAP3
Gene Accession No. Q9Y2W1
Gene Alias TRAP150, thyroid hormone receptor-associated protein 3, thyroid hormone receptor-associated protein complex 150 kDa component, thyroid hormone receptor-associated protein, 150 kDa subunit
Gene Symbols THRAP3
Host Species Rabbit
Immunogen Between 375 and 425
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9967
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.