Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ THRAP5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540517
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540517 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540517 Supplier Invitrogen™ Supplier No. PA540517
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: RLQFGRAPTL PGSAATLQLD GLARAPGQPK IDHLRRLHLG ACPTEECKAC Sequence homology: Cow: 86%; Dog: 93%; Guinea Pig: 93%; Human: 100%; Mouse: 93%; Pig: 100%; Rat: 93%.

Component of the Mediator complex, a coactivator involved in the regulated transcription of nearly all RNA polymerase II-dependent genes. Mediator functions as a bridge to convey information from gene-specific regulatory proteins to the basal RNA polymerase II transcription machinery. Mediator is recruited to promoters by direct interactions with regulatory proteins and serves as a scaffold for the assembly of a functional preinitiation complex with RNA polymerase II and the general transcription factors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen THRAP5
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MED16
Gene Accession No. Q6PGF3, Q9Y2X0
Gene Alias 95kDa; A630083L04; DRIP92; LOW QUALITY PROTEIN: mediator of RNA polymerase II transcription subunit 16; MED16; Mediator complex subunit 16; mediator of RNA polymerase II transcription subunit 16; THRAP5; thyroid hormone receptor associated protein 5; thyroid hormone receptor-associated protein 5; thyroid hormone receptor-associated protein 95; thyroid hormone receptor-associated protein complex 95 kDa component; thyroid hormone receptor-associated protein, 95-kD subunit; TRAP95; Vitamin D3 receptor-interacting protein complex 92 kDa component
Gene Symbols MED16
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human THRAP5 (aa 749-798).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10025, 216154
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.