Learn More
Corning™ Thread Adapters
Catalog No. 07201141 Shop All Corning Products
Change view
Click to view available options
Product Type:
A32/A25
A32/A28
A32/A38
A32/A40
A32/A45
A45/A32
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
07-201-141 A32/A40
07-201-138 A32/A25
07-201-139 A32/A28
07-201-140 A32/A38
07-201-143 A32/A45
07-201-144 A45/A32
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 07-201-141 Supplier Corning™ Supplier No. 6874
Add to Cart
Add to Cart
Corning™ Thread Adapters for use with Bottle Top Dispensers
  • Threaded
  • Accessory for Bottle Top Dispenser
TRUSTED_SUSTAINABILITY
Specifications
Product Type A32/A40
For Use With (Application) Quick and precise dispensing of acids, bases and solvents
For Use With (Equipment) Bottle Top Dispenser