Learn More
Description
- Made of beryllium copper
- Comes with lifetime warranty
- Made in the USA.
- 10 in. in length.
Specifications
Specifications
| Product Type | Thread Cleaning Pick |
| Length (English) | 10 in. |
| Diameter (Metric) | 10 mm |
| Quantity | 1 Ea. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
