Learn More
BURKLE INC Brass barrel screw joint, G 2", outer
Supplier:  BURKLE INC 56010100
Encompass_Preferred
Barrel screw joint made of nickel-plated brass for barrel pump stainless steel. Prevents electro-static charge in combination with the Anti-static set. For the safe transfer of flammable and easily flammable liquids. For containers with R 2" steel fine thread inside.
The container connectors make decanting even easier and safer with their more secure connection to barrels.
Catalog No. NC2742205