Learn More
Grainger Threaded Shank Drill 1/4 HSS
Supplier:  Grainger 16W734
Encompass_Preferred
The Grainger 1/16" Threaded Shank Drill Bit is a high-speed steel bit with a 1/16" diameter and 1/4" shank. It has a straight flute design and a 118 degree point angle for drilling into metal, wood, and plastic. The bit features a threaded shank that allows it to be securely fastened into a drill chuck. Made in the USA, this drill bit is ideal for precision drilling applications requiring small diameter holes.
Catalog No. 50-276-3671
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.