Learn More
Grainger Threaded Shank Drill 3/8 HSS
Supplier:  Grainger 16W736
Encompass_Preferred
The Grainger 3/16" Threaded Shank Drill Bit is a high-speed steel bit with a threaded shank for use in hand drills or drill presses. It has a 118 degree point angle and is designed for drilling into metal, wood, and plastics. The overall length is 2-1/2" and it has a 3/16" diameter. This is a general purpose bit suitable for most drilling applications. The threaded shank ensures secure holding in the drill chuck. Made in the USA.
Catalog No. 50-276-4407
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.