Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synthware™ Threaded Sleeve Type Septum
GREENER_CHOICESDP

Catalog No. 31501984 Shop All Kemtech Products
Change view
Click to view available options
Size:
14/20
19/22
24/40
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
31-501-984 14/20
31-501-985 19/22
31-501-986 24/40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 31-501-984 Supplier Synthware™ Supplier No. RS121420
Add to Cart
Add to Cart

Threaded Sleeve Type Septum made from natural white rubber.

  • Threaded Sleeve Stopper.

Specifications

Autoclavable Autoclavable
Sterility Non-sterile
Product Type Sleeve Stopper
Shape Cylindrical
Stopper Number 14/20
Material Rubber
Color White
Quantity 1 Ea.
For Use With (Application) Organic Synthesis
Size 14/20
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.