Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synthware™ Threaded Sleeve Type Septum
GREENER_CHOICESDP

Catalog No. 31501985 Shop All Kemtech Products
Change view
Click to view available options
Size:
14/20
19/22
24/40
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
31-501-985 19/22
31-501-984 14/20
31-501-986 24/40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 31-501-985 Supplier Synthware™ Supplier No. RS121922
Add to Cart
Add to Cart

Threaded Sleeve Type Septum made from natural white rubber.

  • Threaded Sleeve Stopper.

Specifications

Product Type Sleeve Stopper
Sterility Non-sterile
Autoclavable Autoclavable
Stopper Number 19/22
Shape Cylindrical
Color White
Material Rubber
Quantity 1 Ea.
For Use With (Application) Organic Synthesis
Size 19/22
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.