Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synthware™ Threaded Top Conical Reaction Vial
GREENER_CHOICESDP

Catalog No. 31502110 Shop All Kemtech Products
Change view
Click to view available options
Volume (Metric):
0.1 mL
1 mL
3 mL
5 mL
8 mL
Closure Size:
14/10
5/5
7/10
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Closure Size
31-502-110 5 mL 14/10
31-502-106 0.1 mL 5/5
31-502-107 0.1 mL 7/10
31-502-108 1 mL 7/10
31-502-109 3 mL 14/10
31-502-111 8 mL 14/10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 31-502-110 Supplier Synthware™ Supplier No. V131005
Add to Cart
Add to Cart

Heavy walled conical vials designed for high recovery.

Specifications

Color Clear
Material Borosilicate Glass
Sterility Non-sterile
Disposable Single-use
Volume (Metric) 5 mL
Septum Silicone
For Use With (Application) Organic Synthesis
Graduated Yes
Style Heavy-wall
Closure Material PTFE
Closure Type Compression Cap
Quantity 1 Ea.
Closure Size 14/10
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.