Learn More
MilliporeSigma™ Supelco™ Three Layer Disc Septa
Catalog No. 111011956 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Diameter (Metric) Outer:
12.5 mm
Catalog No. Diameter (Metric) Outer
11-101-1956 12.5 mm
1 options
Catalog No. 11-101-1956 Supplier MilliporeSigma™ Supelco™ Supplier No. 20418
Add to Cart
Add to Cart
Feature a soft inner layer of silicone rubber sandwiched between hard outside layers
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Gas Chromatography
Product Type Disc Septum
Liner Material Silicone Rubber
Diameter (English) Outer 1/2 in.
Diameter (Metric) Outer 12.5 mm
No. of Layers 3
Temperature (Metric) Max 200°C
Quantity 12 Pk.