Learn More
Neese Three-Piece Acid Suit
SDP
Catalog No. 19168352G
Change view
Click to view available options
Size:
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-168-352G 4X-Large
19-168-352F 3X-Large
19-168-352D X-Large
19-168-352C Large
19-168-352H 5X-Large
19-168-352E 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-168-352G Supplier Neese Supplier No. 10096552GRN4X
Add to Cart
Add to Cart
This item is not returnable. View return policy
Offers splash protection with a broad range of chemicals
  • Constructed of heavyweight 13 oz. (368g) PVC/polyester knit coated on both sides
  • Jacket is a minimum of 30 in. long (76cm) with adjustments for various sizes
  • Stand-up collar with snaps to allow for detachable hood
  • Double welded seams
  • Storm flap conceals dielectric snaps
  • Flame retardant
Specifications
Color Green
Hood Type Detached
Material Polyester, PVC
Size 4X-Large
Cuff Style Elastic